missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550711
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total MT-ND3 protein.
MT-ND3 Polyclonal antibody specifically detects MT-ND3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifica
MT-ND3 | |
Polyclonal | |
Unconjugated | |
MT-ND3 | |
1.6.5.3; mitochondrially encoded NADH dehydrogenase 3; Mtnd3; mt-Nd3; NADH dehydrogenase subunit 3; NADH3; NADH-ubiquinone oxidoreductase chain 3; ND3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4537 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P03897 | |
MT-ND3 | |
Synthetic peptide corresponding to a region derived from internal residues of human mitochondrially encoded NADH dehydrogenase 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |