missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTHFR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567577
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
MTHFR Polyclonal antibody specifically detects MTHFR in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifica
MTHFR | |
Polyclonal | |
Unconjugated | |
MTHFR | |
5,10-methylenetetrahydrofolate reductase; 5,10-methylenetetrahydrofolate reductase (NADPH); AI323986; Methylenetetrahydrofolate reductase; methylenetetrahydrofolate reductase (NAD(P)H); MTHFR; RP11-56N19.4 | |
Rabbit | |
affinity chromatography | |
RUO | |
17769, 4524 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P42898, Q9WU20 | |
MTHFR | |
Synthesized peptide derived from human MTHFR. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |