Learn More
ratiolab™ Multibox™ Plus Pipette Tip Box
Descrizione
- For use with the Ratiolab™ MuliTray™ refill systems, Multibox™ plus
- With dust-proof hinged and sliding lid
- Completely removable
- Autoclavable at 121°C, 20 min.
Specifica
Specifica
| Autoclavabile | Autoclavable |
| Materiale | Polycarbonate |
| Da utilizzare con (apparecchiatura) | Pipette Tips Up to 1200 μL Volume |