Learn More
WTW™ MultiLab™ Pro 40 IDS S K
Descrizione
The MultiLab™ Pro IDS from Xylem is a modern multi-parameter measuring device with up to four measurement channels.
- Ease of use
- LIMS ready
- GxP compliance
- Compact and functional design
Highlights:
- Measures pH, ORP, conductivity, dissolved oxygen, ISE (coming soon) and turbidity
- Wireless-ready for IDS modules
- Digital sensor recognition
- Simultaneous measurement with four independent measuring channels
- Brilliant touch display
- Preset and definable buffer sets
- QSC calibration
- AutoRead function
- Export to PDF/A, JSON and CSV
- Special touchscreen glass can be operated with gloves and resistant to alcohol and disinfectants
Scope of delivery:
- Device with stand and power supply, SenTix™ 980 (pH), TetraCon™ 925 (COND), FDO™ 925 (DO), SenTix™ ORP-T 900 (ORP), accessories
Specifica
Specifica
| Calibrazione | pH |
| Certificazione/Conformità | CE Certified |
| Canali | 4 |
| Tipo di display | Color TFT |
| Protezione IP | IP42 |
| Measurement Modes | pH, Redox, mV, Conductivity, Oxygen |
| Intervallo (conduttività) | 0.001 μS/cm to 200 μS/cm |
| Intervallo (pH) | 0.000 to 14.000 |
| Risoluzione (ISE) | depending on Sensor |
| Risoluzione (pH) | depending on Sensor |
| Vedi altri risultati |