Learn More
WTW™ MultiLab™ Pro 40 IDS
Codice prodotto. 30929045
Cambia vista
Click to view available options
Canali:
4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Canali
30929045 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30929045 Fornitore WTW™ N. del fornitore 1FD780
 Articolo soggetto a restrizioni
Spedizione stimata: 19-10-2026
Xylem’s MultiLab™ Pro IDS is a modern high-performance multiparameter device with 4 galvanically isolated measuring channels for pH, ORP, mV, conductivity, oxygen, ISE and turbidity measurement.
The MultiLab™ Pro IDS from Xylem is a modern multi-parameter measuring device with up to four measurement channels.
  • Ease of use
  • LIMS ready
  • GxP compliance
  • Compact and functional design
Highlights:
  • Measures pH, ORP, conductivity, dissolved oxygen, ISE (coming soon) and turbidity
  • Wireless-ready for IDS modules
  • Digital sensor recognition
  • Simultaneous measurement with four independent measuring channels
  • Brilliant touch display
  • Preset and definable buffer sets
  • QSC calibration
  • AutoRead function
  • Export to PDF/A, JSON and CSV
  • Special touchscreen glass can be operated with gloves and resistant to alcohol and disinfectants
Scope of delivery:
  • Device with stand and power supply
TRUSTED_SUSTAINABILITY
Specifica
Calibrazione pH
Certificazione/Conformità CE Certified
Canali 4
Tipo di display Color TFT
Protezione IP IP42
Measurement Modes pH, Redox, mV, Conductivity, Oxygen
Intervallo (conduttività) 0.001 μS/cm to 200 μS/cm
Intervallo (pH) 0.000 to 14.000
Risoluzione (ISE) depending on Sensor
Risoluzione (pH) depending on Sensor
Temperature Range (Metric) depending on Sensor
Tipo di funzione registro Manual or Interval
Product Type Multiparameter Laboratory Measuring Device
Tensione 115/230 VAC