Learn More
Scientific Industries SI™ Multiple Sample Starter Set
Descrizione
For use with Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
- 152mm Platform
- Microtube Foam Insert
- Microplate Foam Insert
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Vortex-Genie 2 family of mixers and Vortex-Genie Pulse |
| Include |
|
| Richiede | Platform Attachment 15577315 |