Learn More
MP Biomedicals MULTISURE™ HIV 1/2 Confirmatory Test
Codice prodotto. 31605226 Sfoglia Tutto MP Biomedicals Prodotti
Cambia vista
Click to view available options
Analiti rilevabili:
HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36, HIV-2 gp105
Dimensione della confezione:
20 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Analiti rilevabili Dimensione della confezione
31605226 HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36, HIV-2 gp105 20 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31605226 Fornitore MP Biomedicals N. del fornitore 93030020
 Articolo soggetto a restrizioni
Spedizione stimata: 04-01-2027
Qualitative rapid in vitro immunochromatographic assay to detect and differentiate HIV-1 and HIV-2 antibodies in serum or plasma
  • MP Diagnostics MULTISURE™ HIV 1/2 Confirmatory Test is intended as a rapid confirmatory test, developed to detect and differentiate antibodies specific to HIV-1 gp120, HIV-1 gp41, HIV-1 p24 and HIV-2 gp36, HIV-2 gp105 antigens in human serum or plasma.
  • Confirm and Differentiate HIV-1 and HIV-2
  • Accurate Result
  • Simple Workflow
  • Flexible: Manual or Automatic Reading of Result
  • Wide Temperature Storage (2 °C to 30 °C)
Specifica
Marchio CE CE-IVD
Analiti rilevabili HIV-1 gp120, HIV-1 gp41, HIV-1 p24, HIV-2 gp36, HIV-2 gp105
Tipo HIV 1/2 Confirmatory Test