Learn More
BD Biosciences™ Multitest CD3/CD16+CD56/CD45/CD19 (CE/IVD)
Product Code. 15865998 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15865998 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15865998 Supplier BD Biosciences™ Supplier No. 342416
 Restricted Item
Spedizione stimata: 30-11-2026
Four-color direct immunofluorescence reagent for identifying and determining the percentages of human T, natural killer, and B lymphyocytes in erhythrocyte-lysed whole blood.
  • Four-color direct immunofluorescence reagent for use with a suitably equipped flow cytometer to identify and determine the percentages and absolute counts of mature human T (CD3+), natural killer (NK) (CD3–CD16+CD56+), and B (CD3–CD19+) lymphocytes in erythrocyte-lysed whole blood.
  • When used with BD Trucount™ tubes, absolute counts of these populations can be enumerated from a single tube.
  • BD Multitest CD3/CD16+CD56/CD45/CD19 and BD Trucount tubes can be used with the BD FACS™ Loader, BD FACSVia™ Loader, and BD FACS™ Universal Loader.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the reagent at 2°C–8°C to maintain stability. Do not use after expiration date shown on the label. Do not freeze the reagent or expose it to direct light during storage or during incubation with cells. Keep the reagent vial dry. Do not use the reagent if you observe any changes in appearance. Precipitation or discoloration indicates instability or deterioration.
Includes Trucount™ Tubes
No. of Tests 50 Tests
Quantity 50 Tests
Regulatory Status CE/IVD
Product Type Immunofluorescence Flow Cytometry Test Reagents