Learn More
ZVG™ Multizell™ Tissues
Descrizione
- Ideal choice for when something needs to be quickly wiped up or cleaned
- Absorbent and lint-free tissues can effectively remove both dry and oily dirt
- Cost-effective Multizell™ tissues are made from 100% recycled material
- Ideal for gas stations, car workshops, and even in the healthcare sector - whether it's for elderly and infant care or for cleaning soiled beds
- Lint-free, highly absorbent, and highly resistant to moisture
- Convenient single-sheet dispensing makes the environmentally friendly Multizell™ tissues significantly more economical to use compared to conventional layered pulp
In the zetPutz™ and zetMedica™ range, you will find more lint-free, highly absorbent 1- to 4-ply cleaning and polishing tissues for individual cleaning purposes.
Specifica
Specifica
| Product Type | Wiping Paper |
| Lunghezza (sistema imperiale) | 10.6 in. |
| Lunghezza (sistema metrico) | 27 cm |
| Larghezza (sistema imperiale) | 12.6 in. |
| Larghezza (sistema metrico) | 32 cm |
| Materiale | Recycled Paper |
| Colore | Natural |
| Monouso | Single-use |
| Strato | 4-ply |