Learn More
MYB Antibody, Novus Biologicals™
Codice prodotto. 18612206 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
50 μg
Dimensione della confezione:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18612206 100 μg 100µL
18636155 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18612206 Fornitore Novus Biologicals N. del fornitore NBP298885100ul
Rabbit Polyclonal Antibody
MYB Polyclonal antibody specifically detects MYB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifica
Antigene MYB
Applicazioni Immunohistochemistry, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000
Formulazione 0.2 um filtered solution in PBS
Alias gene Cmyb;c-myb;c-myb_CDS;efg;Transcriptional activator Myb
Specie ospite Rabbit
Immunogeno Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MYB
Metodo di purificazione Antigen and protein A Affinity-purified
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 4602
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG