Learn More
MYH Antibody, Novus Biologicals™
Codice prodotto. 18286208 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
25 μL
Dimensione della confezione:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18286208 0.1 mL 0.10mL
18480992 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18286208 Fornitore Novus Biologicals N. del fornitore NBP187743
Rabbit Polyclonal Antibody
MYH Polyclonal specifically detects MYH in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifica
Antigene MYH
Applicazioni Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato Unconjugated
Diluizione Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20
Formulazione PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias gene CYP2C, EC 3.2.2, EC 3.2.2.-, hMYH, MGC4416, mutY (E. coli) homolog, MutY homolog, mutY homolog (E. coli), Muyth, MYHA/G-specific adenine DNA glycosylase
Simboli geni MUTYH
Specie ospite Rabbit
Immunogeno This antibody was developed against Recombinant Protein corresponding to amino acids:PDVEECAPNTGQCHLCLPPSEPWDQTLGVVNFPRKASRKPPREESSATCVLEQPGALGAQILLVQRPNSGLLAGLWEFPSVTWEPSEQLQRKALLQELQRWA
Metodo di purificazione Affinity Purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Base Excision Repair, Cancer, DNA Repair
Primario o secondario Primary
ID gene (immissione) 4595
Test di specificità Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
For Research Use Only