missing translation for 'onlineSavingsMsg'
Learn More

Myoferlin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 19548-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899084

  • 121.38€ / 20µL

Spedizione stimata: 11-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The antibody can recognize all the 8 isoforms of MYOF.

Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined.
Specifica

Specifica

Myoferlin
Polyclonal
Unconjugated
MYOF
Fer 1 like protein 3, FER1L3, KIAA1207, MYOF, Myoferlin
Rabbit
Antigen Affinity Chromatography
RUO
26509
-20°C
Liquid
Immunofluorescence, Flow Cytometry, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NZM1
MYOF
Peptide
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali