Learn More
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Codice prodotto. 17244243 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Product Type:
12 V Adapter
Centrifuga da banco
Transport Box with Inserts
Dimensione della confezione:
Pezzo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Dimensione della confezione
17244243 Transport Box with Inserts Pezzo
15334204 Centrifuga da banco Pezzo
17254243 12 V Adapter Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 17244243 Fornitore Thermo Scientific™ N. del fornitore 75004086
  • ONLINE EXCLUSIVE PRICE
    163.00€
    132.30€ / pezzi 1
    Risparmia 30.70€
    18% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 02-02-2027
Our most compact centrifuge, the mySPIN 6 mini centrifuge, is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microcentrifuge tubes.
Bring efficiency and user comfort to your lab stations with the Thermo Scientific mySPIN 6 mini centrifuge. Ergonomic design, tool-free rotors, and quick-spin options make it fast and easy to use. Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL) for customized use.
Enjoy convenience at your personal workstation with our most compact centrifuges, featuring:
  • Small footprint for efficient use of lab space
  • Easy operation and palm-shaped lid for left- or right-handed users
  • Quick-spin feature
  • Quiet operation, low vibration
  • Safety features including unit will not operate without lid in place for safety
  • Includes 2 rotors to accommodate 0.2, 0.5, 1.5, 2.0 mL microcentrifuge tubes and PCR strip tubes
  • Tool-free, quick rotor exchange
TRUSTED_SUSTAINABILITY
Specifica
Include Transport box with inserts
Dimensioni (A P Diam.) 209 x 426 x 383 mm
Product Line mySpin 6
Product Type Transport Box with Inserts