missing translation for 'onlineSavingsMsg'
Learn More

N-(2,6-Dichlorophenyl)-2-fluorobenzamide, 97%, Thermo Scientific™

Catalog Number p-7040011
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
304884-73-9
C13H8Cl2FNO
284.111
MFCD00578380
CSQKIRJRBCAPIY-UHFFFAOYSA-N
n-2,6-dichlorophenyl-2-fluorobenzamide, n-2,6-dichlorophenyl 2-fluorophenyl carboxamide
4372431
N-(2,6-dichlorophenyl)-2-fluorobenzamide
C1=CC=C(C(=C1)C(=O)NC2=C(C=CC=C2Cl)Cl)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
304884-73-9
C13H8Cl2FNO
284.111
MFCD00578380
CSQKIRJRBCAPIY-UHFFFAOYSA-N
n-2,6-dichlorophenyl-2-fluorobenzamide, n-2,6-dichlorophenyl 2-fluorophenyl carboxamide
4372431
N-(2,6-dichlorophenyl)-2-fluorobenzamide
C1=CC=C(C(=C1)C(=O)NC2=C(C=CC=C2Cl)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N-(2,6-Dichlorophenyl)-2-fluorobenzamide
Quantità 250 mg
Peso formulazione 284.12
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'