Learn More
N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]furan-2-carboxamide Hydrochloride, Mikromol™
Descrizione
N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]furan-2-carboxamide Hydrochloride
Specifica
Specifica
| Materiale o nome chimico | 2,3,4,5-Tetradehydro Alfuzosin hydrochloride |
| CAS | 98902-29-5 |
| Nomi componenti o analita | 2,3,4,5-Tetradehydro Alfuzosin hydrochloride |
| Shipping Condition | Room Temperature |
| Formula molecolare | C19 H23 N5 O4 . Cl H |
| Certificazione/Conformità | ISO 9001 |
| Sinonimo | 2-Furancarboxamide, N-[3-[(4-amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]-, hydrochloride (1:1), 2-Furancarboxamide, N-[3-[(4-amino-6,7-dimethoxy-2-quinazolinyl)methylamino]propyl]-, monohydrochloride (9CI), N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]furan-2-carboxamide hydrochloride, 2,3,4,5-Tetradehydroalfuzosin hydrochloride, Alfuzosin Hydrochloride Impurity A as Hydrochloride, Alfuzosin Hydrochloride Impurity A as Hydrochloride, Alfuzosin Impurity A as Hydrochloride, Alfuzosin Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C19H23N5O4.ClH/c1-24(8-5-7-21-18(25)14-6-4-9-28-14)19-22-13-11-16(27-3)15(26-2)10-12(13)17(20)23-19;/h4,6,9-11H,5,7-8H2,1-3H3,(H,21,25)(H2,20,22,23);1H |
| SMILES | Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC(=O)c3occc3 |
| IUPAC Name | N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]furan-2-carboxamide;hydrochloride |
| Vedi altri risultati |