Learn More
LNI Swissgas N.3 Refill for Desiccant Column
Product Code. 17852029 Shop All LNI Swissgas Products
Change view
Click to view available options
Product Type:
Desiccant Column Refill
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17852029 Desiccant Column Refill Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17852029 Supplier LNI Swissgas Supplier No. 6930.00.093
  • ONLINE EXCLUSIVE PRICE
    157.00€
    141.96€ / pezzi 1
    Risparmia 15.04€
    9% Sconto
 Restricted Item
Spedizione stimata: 27-11-2026
N.3 refill for desiccant column.
  • For hydrogen range
  • Replace every 6 months
Specifications
Product Type Desiccant Column Refill