Learn More
N-[[4-[[(4-Amino[1,1'-biphenyl]-3-yl)amino]carbonyl]phenyl]methyl]carbamic Acid Methyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | N-[(4-[((4-Amino[1,1'-biphenyl]-3-yl)amino)carbonyl]phenyl)methyl]carbamic Acid Methyl Ester |
| CAS | 1013330-79-4 |
| Formula molecolare | C22H21N3O3 |
| Quantità | 5 mg |
| InChI Formula | InChI=1S/C22H21N3O3/c1-28-22(27)24-14-15-7-9-17(10-8-15)21(26)25-20-13-18(11-12-19(20)23)16-5-3-2-4-6-16/h2-13H,14,23H2,1H3,(H,24,27)(H,25,26) |
| SMILES | COC(=O)NCc1ccc(cc1)C(=O)Nc2cc(ccc2N)c3ccccc3 |
| IUPAC Name | methyl N-[[4-[(2-amino-5-phenylphenyl)carbamoyl]phenyl]methyl]carbamate |
| Molecular Weight (g/mol) | 375.42 |