missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Bromo-3-methylphenyl)-2-methoxybenzamide, 97%, Thermo Scientific™

Catalog Number 15446058
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
586394-64-1
C15H14BrNO2
320.186
MFCD03241922
DAJGBNVYZLWLSV-UHFFFAOYSA-N
n-4-bromo-3-methylphenyl-2-methoxybenzamide, cambridge id 5937162
2877851
N-(4-bromo-3-methylphenyl)-2-methoxybenzamide
CC1=C(C=CC(=C1)NC(=O)C2=CC=CC=C2OC)Br
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
586394-64-1
C15H14BrNO2
320.186
MFCD03241922
DAJGBNVYZLWLSV-UHFFFAOYSA-N
n-4-bromo-3-methylphenyl-2-methoxybenzamide, cambridge id 5937162
2877851
N-(4-bromo-3-methylphenyl)-2-methoxybenzamide
CC1=C(C=CC(=C1)NC(=O)C2=CC=CC=C2OC)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N-(4-Bromo-3-methylphenyl)-2-methoxybenzamide
Quantità 1 g
Peso formulazione 320.19
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'