Learn More
N-(4-Methoxy-2-methyl-6-quinolinyl)acetamide, TRC
Product Code. 30473932
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30473932 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30473932 Supplier Toronto Research Chemicals Supplier No. TRCM1007951G
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(4-Methoxy-2-methyl-6-quinolinyl)acetamide
CAS 100795-23-1
Molecular Formula C13H14N2O2
Quantity 1 g
Synonym 6-Acetamido-4-methoxy-quinaldine
InChI Formula InChI=1S/C13H14N2O2/c1-8-6-13(17-3)11-7-10(15-9(2)16)4-5-12(11)14-8/h4-7H,1-3H3,(H,15,16)
SMILES COc1cc(C)nc2ccc(NC(=O)C)cc12
Molecular Weight (g/mol) 230.26
Formula Weight 230.1055