Learn More
N-(4-Methyl-2-thiazolyl)-guanidine hydrochloride, TRC
Product Code. 30476891
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476891 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30476891 Supplier Toronto Research Chemicals Supplier No. TRCM33143550MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(4-Methyl-2-thiazolyl)-guanidine hydrochloride
CAS 100599-91-5
Molecular Formula C5H8N4S . HCl
Quantity 50 mg
Synonym Guanidine, N-(4-methyl-2-thiazolyl)-, hydrochloride (1:1) (ACI), Guanidine, (4-methyl-2-thiazolyl)-, monohydrochloride (9CI), 1-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride, N-(4-Methyl-2-thiazolyl)guanidine hydrochloride, (4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride, (4-Methyl-thiazol-2-yl)-guanidine hydrochloride
InChI Formula InChI=1S/C5H8N4S.ClH/c1-3-2-10-5(8-3)9-4(6)7;/h2H,1H3,(H4,6,7,8,9);1H
SMILES Cl.Cc1csc(NC(=N)N)n1
IUPAC Name 2-(4-methyl-1,3-thiazol-2-yl)guanidine;hydrochloride
Molecular Weight (g/mol) 192.67
Formula Weight 192.0236