missing translation for 'onlineSavingsMsg'
Learn More

N,5-Dimethyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 97%, Thermo Scientific™

Catalog Number p-3629719
Click to view available options
Quantità:
250 mg
1 g
Imballaggio:
Amber Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1031843-22-7
C12H15N3
201.27
MFCD11109316
PYMKNYBFTGMBJN-UHFFFAOYSA-N
methyl 5-methyl-1-phenylpyrazol-4-yl methyl amine, 4-methyl aminomethyl-5-methyl-1-phenyl-1h-pyrazole, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methylamine, methyl 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, n,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethyl amine, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine
33589450
N-methyl-1-(5-methyl-1-phenylpyrazol-4-yl)methanamine
CNCC1=C(C)N(N=C1)C1=CC=CC=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
1031843-22-7
C12H15N3
201.27
MFCD11109316
PYMKNYBFTGMBJN-UHFFFAOYSA-N
methyl 5-methyl-1-phenylpyrazol-4-yl methyl amine, 4-methyl aminomethyl-5-methyl-1-phenyl-1h-pyrazole, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methylamine, methyl 5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine, n,5-dimethyl-1-phenyl-1h-pyrazol-4-ylmethyl amine, n-methyl-1-5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine
33589450
N-methyl-1-(5-methyl-1-phenylpyrazol-4-yl)methanamine
CNCC1=C(C)N(N=C1)C1=CC=CC=C1
Materiale o nome chimico N, 5-Dimethyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine
CAS min. % 96.5
CAS max % 100.0
Intervallo percentuale saggio 97%
Imballaggio Amber Glass Bottle
Quantità 250 mg
Peso formulazione 201.27
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'