Learn More
N-Benzyl-4-carboethoxypiperidine, TRC
Specifica
Specifica
| Materiale o nome chimico | N-Benzyl-4-carboethoxypiperidine |
| CAS | 24228-40-8 |
| Formula molecolare | C15 H21 N O2 |
| Quantità | 25 g |
| Sinonimo | 4-Piperidinecarboxylic acid, 1-(phenylmethyl)-, ethyl ester, Isonipecotic acid, 1-benzyl-, ethyl ester (8CI), Ethyl 1-(phenylmethyl)-4-piperidinecarboxylate, 1-(Phenylmethyl)piperidine-4-carboxylic acid ethyl ester, 1-Benzyl-4-carboethoxypiperidine, 1-Benzyl-piperidine-4-carboxylic acid ethyl ester, Ethyl 1-benzylisonipecotate, Ethyl 1-benzylpiperidine-4-carboxylate, Ethyl N-benzylisonipecotate, Ethyl N-benzylisonipecotinate |
| InChI Formula | InChI=1S/C15H21NO2/c1-2-18-15(17)14-8-10-16(11-9-14)12-13-6-4-3-5-7-13/h3-7,14H,2,8-12H2,1H3 |
| SMILES | CCOC(=O)C1CCN(Cc2ccccc2)CC1 |
| IUPAC Name | ethyl 1-benzylpiperidine-4-carboxylate |
| Molecular Weight (g/mol) | 247.33 |
| Peso formulazione | 247.1572 |