Learn More
N-Benzyloxycarbonyl-L-methionine, 98+%, Thermo Scientific™
Product Code. 15468889
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468889 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15468889 Supplier Thermo Scientific Alfa Aesar Supplier No. L08840.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1152-62-1
Melting Point 67°C to 71°C
Molecular Formula C13H17NO4S
MDL Number MFCD00065124
Quantity 5 g
Beilstein 2058696
Synonym z-met-oh, n-cbz-l-methionine, cbz-l-methionine, cbz-met-oh, z-l-methionine, n-carbobenzyloxy-l-methionine, n-carbobenzoxy-l-methionine, benzyloxycarbonyl-l-methionine, n-benzyloxy carbonyl-l-methionine, n-benzyloxycarbonyl-l-methionine
InChI Key FPKHNNQXKZMOJJ-UHFFFAOYNA-N
Optical Rotation −26° (c=1 in Methanol)
SMILES CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
IUPAC Name (2S)-4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
Molecular Weight (g/mol) 283.34
PubChem CID 1550857
Formula Weight 283.35
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-L-methionine