missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-1,3-diaminopropane, 97%, Thermo Scientific Chemicals

Catalog Number p-4258785
Click to view available options
Quantità:
1 mL
5 mL
Imballaggio:
Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
75178-96-0
C8H19N2O2
175.25
MFCD00210021
POHWAQLZBIMPRN-UHFFFAOYSA-O
n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
2735700
tert-butyl N-(3-aminopropyl)carbamate
CC(C)(C)OC(=O)NCCC[NH3+]
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
75178-96-0
C8H19N2O2
175.25
MFCD00210021
POHWAQLZBIMPRN-UHFFFAOYSA-O
n-boc-1,3-propanediamine, n-boc-1,3-diaminopropane, tert-butyl n-3-aminopropyl carbamate, tert-butyl 3-aminopropyl carbamate, n-3-aminopropyl carbamic acid tert-butyl ester, tert-butyl 3-aminopropylcarbamate, 1-boc-amino-1,3-propanediamine, 3-amino-propyl-carbamic acid tert-butyl ester, n-t-boc-1,3-propanediamine, n-tert-butoxycarbonyl-1,3-diaminopropane
2735700
tert-butyl N-(3-aminopropyl)carbamate
CC(C)(C)OC(=O)NCCC[NH3+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N-BOC-1, 3-diaminopropane
Punto di fusione 22.0°C
Punti di ebollizione 94.0°C to 96.0°C (0.2 Torr)
CAS min. % 96.0
CAS max % 100.0
Colore Colorless
Intervallo percentuale saggio 96% min. (GC)
Spettro a infrarossi Authentic
Formula lineare (CH3)3CHOC(O)NH(CH2)3NH2
Imballaggio Glass Bottle
Peso formulazione 174.24
Percent Purity 97%
Densità 0.9980g/mL
Quantità 1 mL
Gravità specifica 0.998
Forma fisica Liquid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'