Learn More
N-Boc-3-hydroxy-L-valine, TRC
Specifica
Specifica
| Chemical Name or Material | N-Boc-3-hydroxy-L-valine |
| CAS | 102507-13-1 |
| Molecular Formula | C10H19NO5 |
| Quantity | 50 mg |
| Synonym | N-[(1,1-Dimethylethoxy)carbonyl]-3-methyl-L-threonine, N-[(1,1-Dimethylethoxy)carbonyl]-3-hydroxy-L-valine, (S)-2-(tert-Butoxycarbonylamino)-3-hydroxy-3-methylbutanoic Acid |
| InChI Formula | InChI=1S/C10H19NO5/c1-9(2,3)16-8(14)11-6(7(12)13)10(4,5)15/h6,15H,1-5H3,(H,11,14)(H,12,13)/t6-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@H](C(=O)O)C(C)(C)O |
| IUPAC Name | (2S)-3-hydroxy-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
| Molecular Weight (g/mol) | 233.26 |
| Formula Weight | 233.1263 |