Learn More
N-Carbobenzyloxy-D-serine-Beta-lactone, TRC
Codice prodotto. 30368604
Cambia vista
Click to view available options
Quantità:
50 mg
100 mg
500 mg
Dimensione della confezione:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30368604 100 mg -
30355175 500 mg 500mg
30353520 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30368604 Fornitore Toronto Research Chemicals N. del fornitore TRCC178400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 98632-91-8
Formula molecolare C11 H11 N O4
Molecular Weight (g/mol) 221.21
Sinonimo Carbamic acid, (2-oxo-3-oxetanyl)-, phenylmethyl ester, (R)-, Carbamic acid, [(3R)-2-oxo-3-oxetanyl]-, phenylmethyl ester (9CI), Phenylmethyl N-[(3R)-2-oxo-3-oxetanyl]carbamate, (R)-Benzyl 2-oxooxetan-3-ylcarbamate
IUPAC Name benzyl N-[(3R)-2-oxooxetan-3-yl]carbamate
SMILES O=C(N[C@@H]1COC1=O)OCc2ccccc2
Specifica
Materiale o nome chimico N-Carbobenzyloxy-D-serine-beta-lactone
InChI Formula InChI=1S/C11H11NO4/c13-10-9(7-15-10)12-11(14)16-6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,14)/t9-/m1/s1
Quantità 100 mg
Peso formulazione 221.0688
Shipping Condition Room Temperature