Learn More
N-Desbenzyl N-tert-butyloxycarbonyl Donepezil, TRC
Codice prodotto. 30367709
Cambia vista
Click to view available options
Quantità:
10 mg
Dimensione della confezione:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30367709 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30367709 Fornitore Toronto Research Chemicals N. del fornitore TRCD28839510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico N-Desbenzyl N-tert-butyloxycarbonyl Donepezil
CAS 948550-60-5
Formula molecolare C22H31NO5
Quantità 10 mg
Sinonimo 4-[(2,3-Dihydro-5,6-dimethoxy-1-oxo-1H-inden-2-yl)methyl]-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C22H31NO5/c1-22(2,3)28-21(25)23-8-6-14(7-9-23)10-16-11-15-12-18(26-4)19(27-5)13-17(15)20(16)24/h12-14,16H,6-11H2,1-5H3
SMILES COc1cc2CC(CC3CCN(CC3)C(=O)OC(C)(C)C)C(=O)c2cc1OC
IUPAC Name tert-butyl 4-[(5,6-dimethoxy-3-oxo-1,2-dihydroinden-2-yl)methyl]piperidine-1-carboxylate
Molecular Weight (g/mol) 389.49
Peso formulazione 389.2202