Learn More
N-Desmethyl-6-O-methylerythromycin (9E)-Oxime, TRC
Product Code. 30007590
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007590 5 mg 5mg
30135446 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30007590 Supplier Toronto Research Chemicals Supplier No. TRCD2923105MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 127182-43-8
Molecular Formula C37 H68 N2 O13
Molecular Weight (g/mol) 748.94
Synonym 3' '-N-Demethyl-6-O-methylerythromycin A (E)-9-oxime, Oxacyclotetradecane, erythromycin deriv., Clarithromycin Imp. M (EP)
IUPAC Name (3R,4S,5S,6R,7R,9R,10E,11S,12R,13S,14R)-14-ethyl-12,13-dihydroxy-10-hydroxyimino-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-6-[(2S,3R,4S,6R)-3-hydroxy-6-methyl-4-(methylamino)oxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecan-2-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@H](NC)[C@H]3O)[C@@](C)(C[C@@H](C)\C(=N/O)\[C@H](C)[C@@H](O)[C@]1(C)O)OC
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C37H68N2O13/c1-14-25-37(10,44)30(41)20(4)27(39-45)18(2)16-36(9,47-13)32(52-34-28(40)24(38-11)15-19(3)48-34)21(5)29(22(6)33(43)50-25)51-26-17-35(8,46-12)31(42)23(7)49-26/h18-26,28-32,34,38,40-42,44-45H,14-17H2,1-13H3/b39-27+/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1
Formula Weight 748.4721
Chemical Name or Material N-Desmethyl-6-O-methylerythromycin A (9E)-Oxime