Learn More
N-Desmethyl Sorafenib, TRC
Product Code. 30349217
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349217 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30349217 Supplier Toronto Research Chemicals Supplier No. TRCD293290100MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Desmethylsorafenib
CAS 284461-74-1
Molecular Formula C21H15ClF3N3O3
Quantity 100 mg
Synonym 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]phenoxy]-2-pyridinecarboxamide, BAY 43-9007, N-[4-Chloro-3-(trifluoromethyl)phenyl]-N’-[4-(2-carbamoyl-4-pyridyloxy)phenyl]urea
InChI Formula InChI=1S/C21H15ClF3N3O3/c22-18-9-6-14(11-17(18)21(23,24)25)28-20(30)27-13-4-7-15(8-5-13)31-16-3-1-2-12(10-16)19(26)29/h1-11H,(H2,26,29)(H2,27,28,30)
SMILES NC(=O)c1cccc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)cc2)c1
IUPAC Name 3-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]phenoxy]benzamide
Molecular Weight (g/mol) 449.81
Formula Weight 449.075