Learn More
N-Desmethyl Terbinafine, TRC
Codice prodotto. 30355275
Cambia vista
Click to view available options
Quantità:
5 mg
Dimensione della confezione:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30355275 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30355275 Fornitore Toronto Research Chemicals N. del fornitore TRCD2943005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico N-Desmethyl Terbinafine
CAS 99473-11-7
Formula molecolare C20 H23 N
Quantità 5 mg
InChI Formula InChI=1S/C20H23N/c1-20(2,3)14-7-4-8-15-21-16-18-12-9-11-17-10-5-6-13-19(17)18/h4-6,8-13,21H,15-16H2,1-3H3/b8-4+
SMILES CC(C)(C)C#C\C=C\CNCc1cccc2ccccc12
IUPAC Name (E)-6,6-dimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine
Molecular Weight (g/mol) 277.4
Peso formulazione 277.183