Learn More
N-Ethyl-2,5-difluorobenzamide, 97%, Thermo Scientific™
Codice prodotto. 15477668
Cambia vista
Click to view available options
Quantità:
250 mg
1 g
Dimensione della confezione:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15477668 250 mg 250mg
15467668 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15477668 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor H61163.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
Este artículo no se puede devolver. Vea la política de devoluciones
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 948717-61-1
Formula molecolare C9H9F2NO
Molecular Weight (g/mol) 185.174
Numero MDL MFCD11843991
InChI Key VMNRGEZTDCWQLU-UHFFFAOYSA-N
Sinonimo benzamide, 2,5-difluoro-n-ethyl
PubChem CID 17683297
IUPAC Name N-ethyl-2,5-difluorobenzamide
SMILES CCNC(=O)C1=C(C=CC(=C1)F)F
Especificaciones
Materiale o nome chimico N-Ethyl-2,5-difluorobenzamide
Quantità 250 mg
Peso formulazione 185.17
Percent Purity 97%