missing translation for 'onlineSavingsMsg'
Learn More

N-Formyl-D-leucine, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7032835
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
44978-39-4
C7H13NO3
159.19
MFCD08275788
HFBHOAHFRNLZGN-MDOHGIEYNA-N
n-alpha-formyl-d-leucine, 2r-2-formamido-4-methylpentanoic acid, r-2-formamido-4-methylpentanoic acid, formyl-d-leucin, n-formyl-d-leucine, d-leucine, n-formyl, r-2-formylamino-4-methylpentanoic acid
13025456
(2R)-2-formamido-4-methylpentanoic acid
CC(C)C[C@@H](NC=O)C(O)=O
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
44978-39-4
C7H13NO3
159.19
MFCD08275788
HFBHOAHFRNLZGN-MDOHGIEYNA-N
n-alpha-formyl-d-leucine, 2r-2-formamido-4-methylpentanoic acid, r-2-formamido-4-methylpentanoic acid, formyl-d-leucin, n-formyl-d-leucine, d-leucine, n-formyl, r-2-formylamino-4-methylpentanoic acid
13025456
(2R)-2-formamido-4-methylpentanoic acid
CC(C)C[C@@H](NC=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N-Formyl-D-leucine
Punto di fusione 132°C to 136°C
Quantità 250 mg
Peso formulazione 159.18
Percent Purity 90%
Grado Technical

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'