missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-[1-(2-furylmethyl)piperid-4-yl]methylamine, 97%, Thermo Scientific™

Catalog Number p-3607874
Click to view available options
Quantità:
250 mg
1 g
Imballaggio:
Amber Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
934570-57-7
C12H20N2O
208.31
MFCD09966159
LISJAFHYRUEIGV-UHFFFAOYSA-N
n-methyl-1-2-furylmethyl piperid-4-yl methylamine, n-1-2-furylmethyl piperidin-4-yl methyl-n-methylamine, 1-furan-2-ylmethyl piperidin-4-yl methyl methyl amine, 1-2-furylmethyl 4-piperidyl methyl methylamine, 1-furan-2-yl methyl piperidin-4-yl methyl methyl amine, 1-1-furan-2-ylmethyl piperidin-4-yl-n-methylmethanamine, 1-1-furan-2-yl methyl piperidin-4-yl-n-methylmethanamine
42556092
CNCC1CCN(CC2=CC=CO2)CC1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
934570-57-7
C12H20N2O
208.31
MFCD09966159
LISJAFHYRUEIGV-UHFFFAOYSA-N
n-methyl-1-2-furylmethyl piperid-4-yl methylamine, n-1-2-furylmethyl piperidin-4-yl methyl-n-methylamine, 1-furan-2-ylmethyl piperidin-4-yl methyl methyl amine, 1-2-furylmethyl 4-piperidyl methyl methylamine, 1-furan-2-yl methyl piperidin-4-yl methyl methyl amine, 1-1-furan-2-ylmethyl piperidin-4-yl-n-methylmethanamine, 1-1-furan-2-yl methyl piperidin-4-yl-n-methylmethanamine
42556092
CNCC1CCN(CC2=CC=CO2)CC1
Materiale o nome chimico N-Methyl-[1-(2-furylmethyl)piperid-4-yl]methylamine
CAS min. % 96.5
CAS max % 100.0
Intervallo percentuale saggio 95%
Imballaggio Amber Glass Bottle
Quantità 250 mg
IUPAC Name 1-[1-(furan-2-ylmethyl)piperidin-4-yl]-N-methylmethanamine
Peso formulazione 208.3
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'