missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methylamine, 90%, Thermo Scientific™

Catalog Number p-3586870
Click to view available options
Quantità:
250 mg
1 g
Imballaggio:
Amber Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
912569-73-4
C13H14F3N3O
285.27
MFCD09817546
AJQKRVBDVZVOBK-UHFFFAOYSA-N
methyl 1-methyl-5-phenoxy-3-trifluoromethyl pyrazol-4-yl methyl amine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine,97+%, n-methyl-1-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methanamine
24229724
N-methyl-1-[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanamine
CNCC1=C(N(N=C1C(F)(F)F)C)OC2=CC=CC=C2
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
912569-73-4
C13H14F3N3O
285.27
MFCD09817546
AJQKRVBDVZVOBK-UHFFFAOYSA-N
methyl 1-methyl-5-phenoxy-3-trifluoromethyl pyrazol-4-yl methyl amine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-1h-pyrazol-4-yl methylamine, n-methyl-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methylamine,97+%, n-methyl-1-1-methyl-5-phenoxy-3-trifluoromethyl-1h-pyrazol-4-yl methanamine
24229724
N-methyl-1-[1-methyl-5-phenoxy-3-(trifluoromethyl)pyrazol-4-yl]methanamine
CNCC1=C(N(N=C1C(F)(F)F)C)OC2=CC=CC=C2
Materiale o nome chimico N-methyl-[1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazol-4-yl]methylamine
CAS min. % 89.5
CAS max % 100.0
Intervallo percentuale saggio 90%
Imballaggio Amber Glass Bottle
Quantità 250 mg
Peso formulazione 285.27
Percent Purity 90%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'