missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[3-(trifluoromethyl)benzyl]amine, 97+%, Thermo Scientific™

Catalog Number p-4510125
Click to view available options
Quantità:
250 mg
1 g
5 g
Imballaggio:
Amber Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
90390-07-1
C9H10F3N
189.181
JFLPPELZYKHKQZ-UHFFFAOYSA-N
n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
485414
N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
CNCC1=CC(=CC=C1)C(F)(F)F
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
90390-07-1
C9H10F3N
189.181
JFLPPELZYKHKQZ-UHFFFAOYSA-N
n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
485414
N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
CNCC1=CC(=CC=C1)C(F)(F)F
Materiale o nome chimico N-Methyl-N-[3-(trifluoromethyl)benzyl]amine
Intervallo percentuale saggio 97%
Imballaggio Amber Glass Bottle
Quantità 250 mg
Peso formulazione 189.18
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'