Learn More
N-Methyl-N-(4-pyridylmethyl)amine, 97+%, Thermo Scientific™
Product Code. 10747834
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10747834 1 g Amber Glass Bottle 1g
10565483 250 mg Amber Glass Bottle 250mg
10597104 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10747834 Supplier Thermo Scientific Maybridge Supplier No. MO00914DA
 Restricted Item
Spedizione stimata: 04-12-2026
Add to basket
Chemical Identifiers
CAS 6971-44-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
InChI Key DNBWGFKLIBQQSL-UHFFFAOYSA-N
Synonym n-methyl-1-pyridin-4-yl methanamine, n-methyl-n-4-pyridylmethyl amine, 4-pyridinemethanamine, n-methyl, methylpyridin-4-ylmethylamine, n-methyl-4-pyridylmethylamine, n-methyl-n-4-pyridinylmethyl amine, methyl pyridin-4-ylmethyl amine, n-methylpyridine-4-methylamine, n-methyl-n-pyridin-4-ylmethyl amine, methyl 4-pyridylmethyl amine
PubChem CID 81436
IUPAC Name N-methyl-1-pyridin-4-ylmethanamine
SMILES CNCC1=CC=NC=C1
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(4-pyridinylmethyl)amine