missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methylformanilide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantità:
10,000 g
250 g
2500 g
500 g
Descrizione
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.
Solubility
Immisible with water.
Notes
Incompatible with oxidizing agents.
N-Methylformanilide is used as a formylating reagent for certain organometallics. It is in combination with phosphoryl chloride used for Vilsmeier-Haack reactions and in heterocycle syntheses. Further, it acts as a swelling agent in the dyeing process of meta-aramid fibers.
Solubility
Immisible with water.
Notes
Incompatible with oxidizing agents.
Specifica
Materiale o nome chimico | N-Methylformanilide |
Punto di fusione | 9°C to 13°C |
Densità | 1.095 |
Punti di ebollizione | 242°C to 244°C |
Punto d'infiammabilità | 126°C (258°F) |
Indice di rifrazione | 1.561 |
Quantità | 250 g |
Beilstein | 636496 |
Informazioni di solubilità | Immisible with water. |
Peso formulazione | 135.17 |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'