Learn More
N,N,beta-Trimethyl-phenethylamine Hydrochloride, TRC
Specifica
Specifica
| Materiale o nome chimico | N,N,beta-Trimethyl-phenethylamine Hydrochloride |
| CAS | 861007-66-1 |
| Formula molecolare | C11H18ClN |
| Quantità | 10 g |
| Sinonimo | N,N,β-Trimethyl-benzeneethanamine Hydrochloride |
| InChI Formula | InChI=1S/C11H17N.ClH/c1-10(9-12(2)3)11-7-5-4-6-8-11;/h4-8,10H,9H2,1-3H3;1H |
| SMILES | CN(C)CC(C)C1=CC=CC=C1.Cl |
| Molecular Weight (g/mol) | 199.72 |
| Peso formulazione | 199.1128 |