missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog Number p-7024630
Click to view available options
Quantità:
100 g
500 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100-36-7
C6H16N2
116.208
MFCD00008176
UDGSVBYJWHOHNN-UHFFFAOYSA-N
n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine
60993
N',N'-diethylethane-1,2-diamine
CCN(CC)CCN
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
100-36-7
C6H16N2
116.208
MFCD00008176
UDGSVBYJWHOHNN-UHFFFAOYSA-N
n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine
60993
N',N'-diethylethane-1,2-diamine
CCN(CC)CCN

N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides. It is also used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides. It is also used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

Solubility
Miscible in water.

Notes
Air sensitive. Store away from oxidizing agents. Protect against electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico N,N-Diethylethylenediamine
Punto di fusione <-70°C
Densità 0.825
Punti di ebollizione 145°C to 147°C
Punto d'infiammabilità 30°C (86°F)
Indice di rifrazione 1.436
Quantità 100 g
Numero UN UN2685
Beilstein 605299
Informazioni di solubilità Miscible in water.
Peso formulazione 116.21
Percent Purity 99%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'