missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropyl-3-methylbenzamide, 97%, Thermo Scientific™

Catalog Number 15446438
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5448-36-2
C14H21NO
219.328
MFCD00587389
WPFBQNDWEOVSGC-UHFFFAOYSA-N
n,n-diisopropyl-3-methylbenzamide, 3-methyl-n,n-di propan-2-yl benzamide, 3-methyl-n,n-bis isopropyl benzamide, cbmicro_015287, cambridge id 5267036, n,n-bis methylethyl 3-methylphenyl carboxamide
226948
3-methyl-N,N-di(propan-2-yl)benzamide
CC1=CC=CC(=C1)C(=O)N(C(C)C)C(C)C
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5448-36-2
C14H21NO
219.328
MFCD00587389
WPFBQNDWEOVSGC-UHFFFAOYSA-N
n,n-diisopropyl-3-methylbenzamide, 3-methyl-n,n-di propan-2-yl benzamide, 3-methyl-n,n-bis isopropyl benzamide, cbmicro_015287, cambridge id 5267036, n,n-bis methylethyl 3-methylphenyl carboxamide
226948
3-methyl-N,N-di(propan-2-yl)benzamide
CC1=CC=CC(=C1)C(=O)N(C(C)C)C(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N,N-Diisopropyl-3-methylbenzamide
Quantità 1 g
Peso formulazione 219.33
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'