Learn More
N',N4-Bis(benzyloxycarbonyl)spermidine, TRC
Specifica
Specifica
| Materiale o nome chimico | N',N4-Bis(benzyloxycarbonyl)spermidine |
| CAS | 97628-87-0 |
| Formula molecolare | C23H31N3O4 |
| Quantità | 25 mg |
| Sinonimo | N-(4-Aminobutyl)-N-[3-[[(phenylmethoxy)carbonyl]amino]propyl]-carbamic Acid Phenylmethyl Ester, (4-Aminobutyl)[3-[[(phenylmethoxy)carbonyl]amino]propyl]-carbamic Acid Phenylmethyl Ester |
| InChI Formula | InChI=1S/C23H31N3O4/c24-14-7-8-16-26(23(28)30-19-21-12-5-2-6-13-21)17-9-15-25-22(27)29-18-20-10-3-1-4-11-20/h1-6,10-13H,7-9,14-19,24H2,(H,25,27) |
| SMILES | NCCCCN(CCCNC(=O)OCc1ccccc1)C(=O)OCc2ccccc2 |
| IUPAC Name | benzyl N-(4-aminobutyl)-N-[3-(phenylmethoxycarbonylamino)propyl]carbamate |
| Molecular Weight (g/mol) | 413.51 |