missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Oleoylethanolamide-d4, TRC
Specifica
Specifica
| Materiale o nome chimico | Oleoyl Ethanolamide-d4 |
| CAS | 946524-36-3 |
| Formula molecolare | C20 D4 H35 N O2 |
| Quantità | 25 mg |
| Sinonimo | (9Z)-N-(2-Hydroxyethyl-1,1,2,2-d4)-9-octadecenamide, (Z)-N-(2-Hhydroxyethyl-d4)-9-octadecenamide, N-(2-Hydroxyethyl-d4)-oleamide, AM 3101, N-(2-Hydroxyethyl-d4)-9-Z-octadecenamide, N-(2-Hydroxyethyl-d4)oleamide, N-Oleoyl-2-aminoethanol-d4, N-Oleoylethanol-d4-amide, N-Oleoylethanol-d4-amine, Oleamide MEA-d4, Oleic Acid Ethanol-d4-amide, Oleic Acid Monoethanol-d4-amide, Oleoylethanol-d4-amide |
| InChI Formula | InChI=1S/C20H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h9-10,22H,2-8,11-19H2,1H3,(H,21,23)/b10-9-/i18D2,19D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])NC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC Name | (Z)-N-(1,1,2,2-tetradeuterio-2-hydroxyethyl)octadec-9-enamide |
| Molecular Weight (g/mol) | 329.554 |
| Peso formulazione | 329.323 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?