Learn More
N-Phenylmethanesulfonamide, TRC
Specifica
Specifica
| Chemical Name or Material | N-Phenylmethanesulfonamide |
| CAS | 1197-22-4 |
| Molecular Formula | C7 H9 N O2 S |
| Quantity | 500 mg |
| Synonym | Methanesulfonamide','N-phenyl-, Methanesulfonanilide (6CI,7CI,8CI), N-Phenylmethanesulfonamide, Methanesulfanilide, Methylsulfonanilide, N-(Methylsulfonyl)aniline, N-Mesylaniline, NSC 11329 |
| InChI Formula | InChI=1S/C7H9NO2S/c1-11(9,10)8-7-5-3-2-4-6-7/h2-6,8H,1H3 |
| SMILES | CS(=O)(=O)Nc1ccccc1 |
| IUPAC Name | N-phenylmethanesulfonamide |
| Molecular Weight (g/mol) | 171.217 |
| Formula Weight | 171.0354 |