Learn More
N-Succinimidyl 4-Azidosalicylate, TRC
Codice prodotto. 30420492
Cambia vista
Click to view available options
Quantità:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30420492 50 mg
30409373 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30420492 Fornitore Toronto Research Chemicals N. del fornitore TRCS69013050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 96602-46-9
Formula molecolare C11 H8 N4 O5
Molecular Weight (g/mol) 276.2
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-azido-2-hydroxybenzoate
SMILES Oc1cc(ccc1C(=O)ON2C(=O)CCC2=O)N=[N+]=[N-]
Specifica
Materiale o nome chimico N-Succinimidyl 4-Azidosalicylate
Quantità 50 mg
InChI Formula InChI=1S/C11H8N4O5/c12-14-13-6-1-2-7(8(16)5-6)11(19)20-15-9(17)3-4-10(15)18/h1-2,5,16H,3-4H2
Peso formulazione 276.0495