missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)ethanolamine, 98%, Thermo Scientific Chemicals

Catalog Number p-78426
Click to view available options
Quantità:
5 mL
25 mL
Imballaggio:
Glass Bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
26690-80-2
C7H15NO3
161.2
MFCD00056657
GPTXCAZYUMDUMN-UHFFFAOYSA-N
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
2733206
tert-butyl N-(2-hydroxyethyl)carbamate
CC(C)(C)OC(=O)NCCO
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
26690-80-2
C7H15NO3
161.2
MFCD00056657
GPTXCAZYUMDUMN-UHFFFAOYSA-N
n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
2733206
tert-butyl N-(2-hydroxyethyl)carbamate
CC(C)(C)OC(=O)NCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico N-(tert-Butoxycarbonyl)ethanolamine
Nota nome 98%
CAS min. % 97.5
CAS max % 100.0
Colore Colorless to Yellow
Intervallo percentuale saggio 97.5% min. (GC)
Spettro a infrarossi Authentic
Formula lineare HOCH2CH2NHCO2C(CH3)3
Imballaggio Glass Bottle
Indice di rifrazione 1.4485 to 1.4505
Peso formulazione 161.2
Percent Purity 98%
Densità 1.0420g/mL
Quantità 5 mL
Punto d'infiammabilità >110°C
Gravità specifica 1.042
Forma fisica Viscous Liquid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'