Learn More
N1-(2-Chlorophenyl)-2-bromoacetamide, 95%, Thermo Scientific™
Product Code. 10191032
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10191032 1 g 1g
10101042 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10191032 Supplier Thermo Scientific Maybridge Supplier No. SPB05477DA
 Restricted Item
Spedizione stimata: 07-12-2026
Add to basket
Chemical Identifiers
CAS 5439-11-2
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.50
MDL Number MFCD00173815
InChI Key WIOJXHICOOQVAB-UHFFFAOYSA-N
Synonym 2-bromo-n-2-chlorophenyl acetamide, n1-2-chlorophenyl-2-bromoacetamide, n-bromoacetyl-2'-chloroacetaniline, wiojxhicooqvab-uhfffaoysa-n, 2-bromo-n-2-chloro-phenyl-acetamide, acetamide, 2-bromo-n-2-chlorophenyl
PubChem CID 225525
Specifications
CAS Min % 97.0
Assay Percent Range 95%
Linear Formula ClC6H4NHC(O)CH2Br
Quantity 1 g
Formula Weight 248.51
Percent Purity 95%
Chemical Name or Material N1-(2-chlorophenyl)-2-bromoacetamide