Learn More
N6-Trifluoroacetyl-L-lysine N-Carboxyanhydride, TRC
Specifica
Specifica
| Materiale o nome chimico | N6-Trifluoroacetyl-L-lysine N-Carboxyanhydride |
| CAS | 42267-27-6 |
| Formula molecolare | C9 H11 F3 N2 O4 |
| Quantità | 10 g |
| Sinonimo | Acetamide','N-[4-[(4S)-2,5-dioxo-4-oxazolidinyl]butyl]-2,2,2-trifluoro-, Acetamide','N-[4-(2,5-dioxo-4-oxazolidinyl)butyl]-2,2,2-trifluoro-','(S)-, N-[4-[(4S)-2,5-Dioxo-4-oxazolidinyl]butyl]-2,2,2-trifluoroacetamide, N-Trifluoroacetyl-L-lysine N-carboxyanhydride, Nε-Trifluoroacetyl-L-lysine N-carboxyanhydride |
| InChI Formula | InChI=1S/C9H11F3N2O4/c10-9(11,12)7(16)13-4-2-1-3-5-6(15)18-8(17)14-5/h5H,1-4H2,(H,13,16)(H,14,17)/t5-/m0/s1 |
| SMILES | FC(F)(F)C(=O)NCCCC[C@@H]1NC(=O)OC1=O |
| IUPAC Name | N-[4-[(4S)-2,5-dioxo-1,3-oxazolidin-4-yl]butyl]-2,2,2-trifluoroacetamide |
| Molecular Weight (g/mol) | 268.19 |
| Peso formulazione | 268.0671 |