Learn More
Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks
Descrizione
Store, mix and dispense reagents in Thermo Scientific™ Nalgene™ Closed-Dome Polypropylene Tanks, designed for use as closed containment systems. Offered in sizes ranging from 75 to 380L , tanks feature a 150mm neck opening fitted with a gasketed closure for secure sealing.
Features:
- Tanks may be fitted with spigots
- Autoclavable tanks and closures feature polypropylene construction
Compatible with:
Use with Closed-Dome Tank Closure w/ Mixer Support Assembly (Mfr. No. 2651-0200), BioTech Overhead Mixer (Mfr. No. 2653-0010 and 2653-0020) and Lower Assemblies for BioTech Mixers (Mfr. No. 2654-0030, 2654-0055, 2654-0075 & 2654-0100).
Specifica
Specifica
| Certificazione/Conformità | Comply with FDA Reg. 177.1520 and USP Class VI |
| Diametro (sistema imperiale) esterno | 18.5 in. |
| Diametro (sistema metrico) esterno | 47 cm |
| Tipo | Closed-dome Tank |
| Materiale | Polypropylene |
| Capacità (sistema imperiale) | 30 gal. |
| Capacità (sistema metrico) | 115 L |
| Altezza (sistema imperiale) | 38.625 in. |
| Altezza (sistema metrico) | 98.1 cm |
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.