Learn More
Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure: Bulk Pack
Descrizione
Store samples and ship small volumes of valuable reagents and light-sensitive diagnostic samples with Thermo Scientific™ Nalgene™ HDPE Diagnostic Bottles without Closure. Safe and leakproof† when used with Nalgene closures, these bottles can also be used in test kits and many other small-volume applications. Bottles are made of opaque white, high-density polyethylene and are packed without closures. They can be matched with various colored closures (Cat. No. 362150 series).
- Nonsterile
- Durable, uniform walls are resistant to splitting or puncturing
- Leakproof† when assembled with Nalgene closure (must be ordered separately)
Ordering Information:
Bottles are bulk packed without closures. 2000 per case. Order colored closures separately (Cat. No. 362150).
Specifica
Capacità | 0.125 oz., 4 mL |
Materiale chiusura | Polypropylene |
Dimensione chiusura | 13-415 mm |
Diametro (sistema metrico) | 16 mm |
Altezza (sistema imperiale) | 1.61 in. |
Altezza (sistema metrico) | 41 mm |
Larghezza (sistema imperiale) | 0.62 in. |
Product Type | Diagnostic Bottle |
Capacità (sistema metrico) | 4 mL |
Capacità (sistema imperiale) | 0.125 oz. |
Vedi altri risultati |
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.