Learn More
Thermo Scientific™ Nalgene™ Lightweight Cylindrical LLDPE Tank with Cover and Spigot
Codice prodotto. 11968457
Cambia vista
Click to view available options
Capacità (sistema metrico):
18.9 L
28.4 L
37.9 L
56.8 L
113.6 L
208 L
Dimensione della confezione:
Pezzo
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) unitSize
11968457 18.9 L Pezzo
11978457 28.4 L Pezzo
10606951 37.9 L Pezzo
11988457 56.8 L Pezzo
10728871 113.6 L Pezzo
11990349 208 L Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 11968457 Fornitore Thermo Scientific™ N. del fornitore 541020005
  • ONLINE EXCLUSIVE PRICE
    244.00€
    230.00€ / pezzi 1
    Risparmia 14.00€
    5% Sconto
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Aggiungi al carrello
An economical alternative to stainless steel, the Thermo Scientific™ Nalgene™ Lightweight Cylindrical LLDPE Tank with Cover and Spigot offers excellent resistance to stress cracking.
An economical alternative to stainless steel, the Thermo Scientific™ Nalgene™ Lightweight Cylindrical LLDPE Tank with Cover and Spigot offers excellent resistance to stress cracking and features a spigot installed for draw off. Ideal for less rigorous applications. Available in sizes ranging from 5 to 55 gal. (19 to 208L).
  • Approximate wall thickness: 0.09 in. (2.4mm)
  • Equipped with Nalgene Needle Spigot (Cat. No. 96423) for dispensing
  • Not autoclavable
Note:
Tanks used at elevated temperatures or with high specific-gravity liquids should be externally supported.
TRUSTED_SUSTAINABILITY
Specifica
Diametro (sistema imperiale) esterno 11 in.
Spessore della parete 2.4 mm
Diametro (sistema metrico) esterno 27.9 cm
Tipo Tank with Cover and Spigot
Materiale LLDPE
Capacità (sistema imperiale) 5 gal.
Capacità (sistema metrico) 18.9 L
Altezza (sistema imperiale) 15 in.
Altezza (sistema metrico) 38.1 cm
N. per custodia 1
N. per confezione 1
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Caution:
Plastic tanks are generally subject to more severe conditions that plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.